Description
- Polyethylene shell and suspension system work together as protection system
- Self-adjusting crown straps for comfortable fit
- Slotted design works with accessories
- Available in standard (61/2 to 8) size
- Four-point webbed Fas-Trac ratchet suspension with vinyl sweatband and large easy-adjustment knob
Order Info
Contact your Fisher Scientific Customer Service Representative for ordering information for sizes not shown.
Certifications
SEI certified.
Compliance
Meet or exceed all applicable requirements of ANSI™ Z89.1-2003 for Type I helmets, Class E and CSA Z94.1 2005.
Specifications
Specifications
| Certifications/Compliance | ANSI Z89.1-2014, CSA Z94.1-2015 |
| Color | Hi-vis Orange |
| Material | Polyethylene |
| Size | Standard |
| Suspension Type | Fas-Trac Ratchet |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.