Learn More
MSA™ V-Gard™ Slotted Protective Cap with Fas-Trac™ Suspension
Catalog No. 189993937 Shop All MSA Products
Change view
Click to view available options
Color:
Black
Blue
Gold
Gray
Green
Hi-vis Lime
Hi-vis Orange
Orange
Red
Robin Egg Blue
White
Yellow
Size:
Large
Small
Standard
Standard (6.5 to 8 in.)
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
18-999-3937 Gold Standard
18-999-3887 White Large
18-999-3934 Gray Standard
19-043-675 White Standard
19-058288 Blue Standard
18-999-3933 Orange Standard
18-999-3936 Hi-vis Orange Standard
19-058280 Yellow Standard
19-716-777 Black Standard (6.5 to 8 in.)
18-999-3886 White Small
18-999-3935 Green Standard
18-999-3932 Red Standard
19-122-849 Hi-vis Lime Standard
19-167-085 Robin Egg Blue Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Not specifically designed to protect against lateral blows from the front, side, or rear.
Catalog No. 18-999-3937 Supplier MSA™ Supplier No. 475365
Add to Cart
Add to Cart
Not specifically designed to protect against lateral blows from the front, side, or rear.
Lightweight for all-day comfort
  • Polyethylene shell and suspension system work together as protection system
  • Self-adjusting crown straps for comfortable fit
  • Slotted design works with accessories
  • Available in standard (61/2 to 8) size
  • Four-point webbed Fas-Trac ratchet suspension with vinyl sweatband and large easy-adjustment knob
Order Info
Contact your Fisher Scientific Customer Service Representative for ordering information for sizes not shown.
Certifications
SEI certified.
Compliance
Meet or exceed all applicable requirements of ANSI™ Z89.1-2003 for Type I helmets, Class E and CSA Z94.1 2005.
Specifications
Certifications/Compliance ANSI Z89.1-2014, CSA Z94.1-2015
Color Gold
Material Polyethylene
Size Standard
Suspension Type Fas-Trac Ratchet
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.