Learn More
MSA™ Welder's Hood
Catalog No. 189993357 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Welder Adapter
Catalog No. For Use With (Equipment)
18-999-3357 Welder Adapter
1 options
This item is not returnable. View return policy
Catalog No. 18-999-3357 Supplier MSA™ Supplier No. 472859
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protects welder's head, neck and shoulders from welding splatter and sparks
  • Rubberized glass yarn hood with rubber seal
  • Fits snugly over welder adapter
  • Clip-on
Specifications
For Use With (Equipment) Welder Adapter
Includes Rubber seal
Material Rubberized Glass Yarn
Quantity 1 Ea.