Learn More
MSA™ Workman™ Self Retracting Lanyards
Catalog No. 17200102 Shop All MSA Products
Change view
Click to view available options
Material:
Stainles Steel Cable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17-200-102 Stainles Steel Cable
17-200-101 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-200-102 Supplier MSA™ Supplier No. 10120724
  • RFID-enabled product for simplified product tracking and inspection
  • Integral, ergonomic carrying handle for easier transport and installation
  • 400 lb. working capacity for increased versatility
  • Swivel 36CS snaphook with load indicator at lifeline end greatly limits twisting of cable; also alerts users that unit has been involved in a fall
  • 30 ft. length for extra mobility
Certifications
CSA Z259.2.2 certified
Compliance
Meets OSHA 1926.502, 1910.66 and ANSI Z359.1-2007
Specifications
Material Stainles Steel Cable
Length (English) 30 ft.
Certifications/Compliance Meets ANSI Z359.1-2007 and OSHA 1926.502, 1910.66. CSA Z259.2.2 certified.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.