Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ XLS Cap Mounted Ear Muff

Catalog No. 19815456 Shop All MSA Products
Change view
Click to view available options
XLS muff for slotted caps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-815-456 XLS muff for slotted caps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-815-456 Supplier MSA™ Supplier No. 10061535

XLS Cap Mounted Ear Muffs are to be used with MSA™ Slotted Hard Hats and provide a high level of comfort in loud environments.

  • Offers protection in low-moderate noise environments
  • Easy-to-remove ear cushions for quick cleaning and replacement
  • Features a spring design for a high level of comfort
  • Choose from three distinct wearing positions

Specifications

Color Black, Red
ANSI S3.1974 Yes
CSA Z94.2 Class A
Cushion Color Black
For Use With (Equipment) MSA™ Slotted Hard Hats
For Use With (Application) General industry, manufacturing, repair and maintenance, construction, oil and gas, forestry, shipbuilding, mining
High Visibility No
Model 10061535
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 23 dB
Product Type XLS
Dielectric No
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.