Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSANTD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558498
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558498 Supplier Invitrogen™ Supplier No. PA558498
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEITISEARC LELHMEIDWI PIAHSKPTGG NVVQYLLPGG IPKSPGLYAI GYEECIERPL SPHMEQSSLD PGKEGRVDLE TLSAQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 91%.

Myb/SANT-like DNA-binding domain-containing protein 2 (MSANTD2) contains 1 myb-like domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSANTD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MSANTD2
Gene Accession No. Q6P1R3
Gene Alias C11orf61; FLJ23342; MSANTD2; Myb/SANT DNA binding domain containing 2; Myb/SANT-like DNA-binding domain containing 2; myb/SANT-like DNA-binding domain-containing protein 2
Gene Symbols MSANTD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MSANTD2. Recombinant protein control fragment (Product #RP-102847).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79684
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.