Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSANTD2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501568443
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8443 Supplier Bethyl Laboratories, Inc Supplier No. A304704A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Myb/SANT-like DNA-binding domain-containing protein 2 (MSANTD2) contains 1 myb-like domain.

Specifications

Antigen MSANTD2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MSANTD2
Gene Accession No. Q6P1R3
Gene Alias C11orf61, chromosome 11 open reading frame 61, FLJ23342, Myb/SANT-like DNA-binding domain containing 2
Gene Symbols MSANTD2
Host Species Rabbit
Immunogen Between 225 and 275
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79684
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.