Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSANTD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571420 Supplier Invitrogen™ Supplier No. PA571420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MSANTD3 Polyclonal Antibody for Western Blot

MSANTD3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include identical protein binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSANTD3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MSANTD3
Gene Accession No. Q96H12
Gene Alias 2410046L14Rik; 2810429P03Rik; 5730528L13Rik; C8H9orf30; C9orf30; L8; MSANTD3; Myb/SANT DNA binding domain containing 3; Myb/SANT-like DNA-binding domain containing 3; myb/SANT-like DNA-binding domain-containing protein 3; Mysantd3; RGD1308165; UPF0439 protein C9orf30; UPF0439 protein C9orf30 homolog
Gene Symbols MSANTD3
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human MSANTD3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91283
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.