Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH2 Antibody (MSH2/6549R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308958
Description
MSH2 Monoclonal specifically detects MSH2 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
MSH2 | |
Monoclonal | |
1 mg/mL | |
Western Blot : 1-2 ug/mL, Immunohistochemistry-Paraffin : 1-2 ug/mL | |
COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2 | |
Recombinant full-length human MSH2 protein | |
100 μg | |
Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors | |
4436 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG κ |
Western Blot, Immunohistochemistry (Paraffin) | |
MSH2/6549R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction