Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH2 Antibody (rMSH2/6548), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308956R
Description
MSH2 Monoclonal antibody specifically detects MSH2 in Human, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
MSH2 | |
Monoclonal | |
DyLight 550 | |
COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2 | |
Recombinant full-length human MSH2 protein (Uniprot: IDP43246) | |
100 μg | |
Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors | |
4436 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin) | |
rMSH2/6548 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Canine, Feline | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction