Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH2 Monoclonal Antibody (OTI10F7), TrueMAB™, OriGene
SDP

Catalog No. 502543396
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3396 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3396 Supplier OriGene Supplier No. CF814210
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MSH2 Monoclonal antibody specifically detects MSH2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen MSH2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI10F7
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene CR2
Gene Accession No. P20023
Gene Alias AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 208-587 of human CD21 (NP_001868) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1380
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.