Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MSH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529160
Description
Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (93%), Rat (92%), Pig (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plays a role in neurogenesis and neuronal migration. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
Specifications
| MSH2 | |
| Polyclonal | |
| Unconjugated | |
| Msh2 | |
| AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4436 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.01 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P43246 | |
| Msh2 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human MSH2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction