Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540136 Supplier Invitrogen™ Supplier No. PA540136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Yeast cells. Predicted reactivity: Yeast (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plays a role in neurogenesis and neuronal migration. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH2
Applications Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Description MSH2 Polyclonal Antibody, 100μL, 0.33mg/mL Concentration, MSH2 Antigen, Recombinant Protein Encompassing a Sequence within the C-terminus Region of Saccharomyces Cerevisiae S288c MSH2 Immunogen, Rabbit Host, Yeast Species Reactivity, Unconjugated, IgG
Dilution 1:500 to 1:3000
Formulation PBS with 20% glycerol and 0.01% Thimerosal; pH 7
Gene Msh2
Gene Accession No. P25847
Gene Alias AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W
Gene Symbols Msh2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of Saccharomyces cerevisiae S288c MSH2. The exact sequence is proprietary.
Positive Control Yeast cells
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 854063
Packaging 1 Each
Includes 0.01% Thimerosal
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.