Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MSH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586911
Description
Purity is > 95% by SDS-PAGE.
Plays a role in neurogenesis and neuronal migration. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
Specifications
MSH2 | |
Polyclonal | |
Unconjugated | |
Msh2 | |
AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4436 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P43246 | |
Msh2 | |
Synthetic peptide, corresponding to amino acids 545-589 of Human MSH2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction