Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSH2 Recombinant Rabbit Monoclonal Antibody (BLR116H)
GREENER_CHOICE

Catalog No. PIMA544423
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544423 100 μL
PIMA544424 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544423 Supplier Invitrogen™ Supplier No. MA544423
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

Plays a role in neurogenesis and neuronal migration. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR116H
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
Gene Msh2
Gene Accession No. P43246
Gene Alias AI788990; COCA1; DNA mismatch repair protein MSH2; FCC1; hMSH2; HNPCC; HNPCC1; LCFS2; mismatch repair ATPase MSH2; mismatch repair protein; MSH2; Msh2p; mutS homolog 2; mutS homolog 2 (E. coli); mutS homolog 2, colon cancer, nonpolyposis type 1; mutS protein homolog 2; O0935; PMS5; YOL090W
Gene Symbols Msh2
Host Species Rabbit
Immunogen Peptide representing a region between residue 625 and 675 of Human DNA mismatch repair protein Msh2.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4436
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.