Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSH3 Recombinant Rabbit Monoclonal Antibody (RM405)
GREENER_CHOICE

Catalog No. PIMA536071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536071 Supplier Invitrogen™ Supplier No. MA536071
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MSH3 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

Component of the post-replicative DNA mismatch repair system (MMR). Heterodimerizes with MSH2 to form MutS beta which binds to DNA mismatches thereby initiating DNA repair. When bound, the MutS beta heterodimer bends the DNA helix and shields approximately 20 base pairs. MutS beta recognizes large insertion-deletion loops (IDL) up to 13 nucleotides long. After mismatch binding, forms a ternary complex with the MutL alpha heterodimer, which is thought to be responsible for directing the downstream MMR events, including strand discrimination, excision, and resynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone RM405
Concentration 0.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene MSH3
Gene Accession No. P20585
Gene Alias D13Em1; Divergent upstream protein; DNA mismatch repair protein Msh3; DUC1; DUG; DUP; hMSH3; Mismatch repair protein 1; MRP1; MSH3; mutS homolog 3; mutS homolog 3 (E. coli); protein repair-1; Protein repair-3; REP-1; Rep3; Rep-3
Gene Symbols MSH3
Host Species Rabbit
Immunogen A peptide corresponding to residues near the N-terminus of human MSH3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4437
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.