Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH3 Polyclonal Antibody
MSH3 Polyclonal Antibody
Supplier: Thermo Scientific PA596387
Description
MSH3 Polyclonal Antibody for Western Blot

Specifications
MSH3 | |
Polyclonal | |
Unconjugated | |
MSH3 | |
D13Em1; Divergent upstream protein; DNA mismatch repair protein Msh3; DUC1; DUG; DUP; hMSH3; Mismatch repair protein 1; MRP1; MSH3; mutS homolog 3; mutS homolog 3 (E. coli); protein repair-1; Protein repair-3; REP-1; Rep3; Rep-3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4437 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20585 | |
MSH3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 617-761 of human MSH3 (NP_0046962). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction