Learn More
Invitrogen™ MSH4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589583
Description
Immunogen sequence: QIMAQIGSYV PAEYSSFRIA KQIFTRISTD DDIETNSSTF MKEMKEIAYI LHNANDKSLI LIDELGRGTN TEEGIGICYA VCEYLLSLKA FTLFATHFLE LCHIDALYPN VENMHFEVQH VKNTSRNKEA ILYTYKLSKG LTEEKNYGLK AAEVSSLPPS IVLDAKEITT QITRQILQNQ RSTPEMERQR AVYHLATRLV QTARNSQLDP DSLRIYLSNL KKKYKEDFPR TEQVPEKTEE; Positive Samples: SKOV3.
MSH4 is involved in meiotic recombination. MSH4 is also required for reciprocal recombination and proper segregation of homologous chromosomes at meiosis.
Specifications
MSH4 | |
Polyclonal | |
Unconjugated | |
MSH4 | |
4930485C04Rik; AV144863; hMSH4; mMsh4; Msh4; mutS homolog 4; mutS homolog 4 (E. coli); MutS homolog 4 variant iota 1; MutS homolog 4 variant theta 1; mutS protein homolog 4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4438 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.99 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15457 | |
MSH4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 697-936 of human MSH4 (NP_0024312). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.