Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115593
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115593 Supplier Invitrogen™ Supplier No. PA5115593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MSH4.

MSH4 is involved in meiotic recombination. MSH4 is also required for reciprocal recombination and proper segregation of homologous chromosomes at meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MSH4
Gene Accession No. O15457, Q99MT2
Gene Alias 4930485C04Rik; AV144863; hMSH4; mMsh4; Msh4; mutS homolog 4; mutS homolog 4 (E. coli); MutS homolog 4 variant iota 1; MutS homolog 4 variant theta 1; mutS protein homolog 4
Gene Symbols MSH4
Host Species Rabbit
Immunogen A synthesized peptide derived from human MSH4(Accession O15457), corresponding to amino acid residues G103-S153.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295541, 4438, 55993
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.