Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541989 Supplier Invitrogen™ Supplier No. PA541989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SSARDTNYPQ TLKTPLSTGN PQRSGYKSWT PQVGYSASSS SAISAHSPSV Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 86%; Rat: 93%.

MSH4 is involved in meiotic recombination. MSH4 is also required for reciprocal recombination and proper segregation of homologous chromosomes at meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MSH4
Gene Accession No. O15457
Gene Alias 4930485C04Rik; AV144863; hMSH4; mMsh4; Msh4; mutS homolog 4; mutS homolog 4 (E. coli); MutS homolog 4 variant iota 1; MutS homolog 4 variant theta 1; mutS protein homolog 4
Gene Symbols MSH4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MSH4 (aa 107-156).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4438
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.