Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH4 Rabbit anti-Plant, Polyclonal, FabGennix

Catalog No. 502207829
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7829 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7829 Supplier Fabgennix Inc Supplier No. MSH4401AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MSH4 Polyclonal antibody specifically detects MSH4 in Plant samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen MSH4
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Alias 4930485C04Rik; AV144863; hMSH4; mMsh4; Msh4; mutS homolog 4; mutS homolog 4 (E. coli); MutS homolog 4 variant iota 1; MutS homolog 4 variant theta 1; mutS protein homolog 4
Host Species Rabbit
Immunogen N-epitope synthetic peptide taken within amino acid region 1-100 MSH4 gene
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.