Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH5 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298819MFV450
Description
MSH5 Polyclonal antibody specifically detects MSH5 in Human samples. It is validated for Western BlotSpecifications
MSH5 | |
Polyclonal | |
Western Blot | |
DKFZp434C1615, G7, hMSH5, MGC2939, mutS (E. coli) homolog 5, mutS homolog 5 (E. coli), mutS protein homolog 5, MUTSH5, NG23 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human MSH5. | |
0.1 mL | |
DNA Repair, Mismatch Repair, modENCODE Antibodies | |
4439 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction