Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541990
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541990 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541990 Supplier Invitrogen™ Supplier No. PA541990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DENMTRFLGK LASQEHREPK RPEIIFLPSV DFGLEISKQR LLSGNYSFIP Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 100%.

MSH5 is a member of the mutS family of proteins that are involved in DNA mismatch repair or meiotic recombination processes. This protein is similar to a Saccharomyces cerevisiae protein that participates in meiotic segregation fidelity and crossing-over. This protein forms heterooligomers with another member of this family, mutS homolog 4. This gene encodes a member of the mutS family of proteins that are involved in DNA mismatch repair or meiotic recombination processes. This protein is similar to a Saccharomyces cerevisiae protein that participates in meiotic segregation fidelity and crossing-over. This protein forms heterooligomers with another member of this family, mutS homolog 4. Alternative splicing results in four transcript variants encoding three different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MSH5
Gene Accession No. O43196
Gene Alias DKFZp434C1615; G7; hMSH5; MGC2939; MSH5; Mut5; mutS homolog 5; mutS homolog 5 (E. coli); mutS protein homolog 5; MUTSH5; NG23
Gene Symbols MSH5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MSH5 (aa 107-156).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4439
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.