Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH6 Antibody (021), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289318MFV500
Description
MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| MSH6 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 021 | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein | |
| This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human MSH6. | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Repair, Mismatch Repair | |
| 2956 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction