Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH6 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100328AF700
Description
MSH6 Polyclonal antibody specifically detects MSH6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MSH6 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein | |
The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human MutS homolog 6 using the numbering given in entry NP_000170.1 (GeneID 2956). | |
0.1 mL | |
Breast Cancer, Cancer, DNA Repair, Mismatch Repair | |
2956 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction