Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSH6 Monoclonal Antibody (3E1)
GREENER_CHOICE

Catalog No. PIMA517129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517129 Supplier Invitrogen™ Supplier No. MA517129
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17129 targets MSH6 in FACS, ICC, IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17129 immunogen is purified recombinant fragment of human MSH6 expressed in E. Coli. MA5-17129 detects MSH6 which has a predicted molecular weight of approximately 160kDa.

The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced malignant transformation and hence the prevention of skin cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH6
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3E1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene MSH6
Gene Accession No. P52701
Gene Alias AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12
Gene Symbols MSH6
Host Species Mouse
Immunogen Purified recombinant fragment of human MSH6 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2956
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.