Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSH6 Monoclonal Antibody (SP93)
GREENER_CHOICE

Catalog No. PIMA516380
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516380 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516380 Supplier Invitrogen™ Supplier No. MA516380
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is rectal carcinoma.

The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced malignant transformation and hence the prevention of skin cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone SP93
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene MSH6
Gene Accession No. P52701
Gene Alias AU044881; AW550279; cb326; DNA mismatch repair protein Msh6; g/T mismatch-binding protein; GTBP; GTMBP; hMSH6; HNPCC5; HSAP; LOC100360342; mismatch repair protein Msh6; Msh6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; mutS homolog 6 (E. coli); MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein; wu:fb36c12
Gene Symbols MSH6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to internal region of human MSH6 protein.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2956
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.