Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH6 Mouse anti-Human, Unconjugated, Clone: MSH6/2111, Abnova™

Catalog No. 89337531 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-531 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-531 Supplier Abnova Corporation Supplier No. MAB21038
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for ELISA, WB-Ce.

Specifications

Antigen MSH6
Applications ELISA, Western Blot
Classification Monoclonal
Clone MSH6/2111
Conjugate Unconjugated
Description mutS homolog 6 (E. coli)
Gene MSH6
Gene Alias GTBP, HNPCC5, HSAP
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 374-540 of human MSH6
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2956
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.