Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSI1 Monoclonal Antibody (2A12)
GREENER_CHOICE

Catalog No. PIMA515820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515820 Supplier Invitrogen™ Supplier No. MA515820
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15820 targets MSI1 in indirect ELISA, FACS and WB applications and shows reactivity with Human samples. The MA5-15820 immunogen is purified recombinant fragment of human MSI1 expressed in E. Coli. MA5-15820 detects MSI1 which has a predicted molecular weight of approximately 39kDa.

Musashi-1 belongs to the Musashi family of RNA-binding proteins which play a role in cell fate determination, and maintenance of stem-cell state. Musashi family proteins are also involved in post transcriptional gene regulation. The gene encodes a protein with two conserved tandem RNA recognition motifs with highly conserved RNP (ribonucleoprotein) motifs. In mammals, Musashi-1 is expressed in fetal kidney, brain, liver and lung, and in adult brain and pancreas. In humans, the gene is located on chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSI1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2A12
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene MSI1
Gene Accession No. O43347
Gene Alias fb40b12; hypothetical protein LOC541389; m-Msi-1; ms1; msi1; msi1b; Msi1h; Musahi1; musashi homolog 1; Musashi homolog 1(Drosophila); musashi RNA binding protein 1; musashi RNA binding protein 1b; musashi RNA-binding protein 1; musashi1; musashi-1; RNA-binding protein Musashi homolog 1; RNA-binding protein Musashi homolog 1 L+35bp; RNA-binding protein Musashi homolog 1 Long; RNA-binding protein Musashi homolog 1b; wu:fb40b12; zgc:103751; zMsi1
Gene Symbols MSI1
Host Species Mouse
Immunogen Purified recombinant fragment of human MSI1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.