Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSI1 Monoclonal Antibody (OTI2G9), TrueMAB™, OriGene
SDP

Catalog No. 502541644
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1644 30 μL
50-167-3374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1644 Supplier OriGene Supplier No. TA502311S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MSI1 Monoclonal antibody specifically detects MSI1 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen MSI1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2G9
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene STRIP1
Gene Accession No. Q5VSL9
Gene Alias fb40b12; hypothetical protein LOC541389; m-Msi-1; ms1; msi1; msi1b; Msi1h; Musahi1; musashi homolog 1; Musashi homolog 1(Drosophila); musashi RNA binding protein 1; musashi RNA binding protein 1b; musashi RNA-binding protein 1; musashi1; musashi-1; RNA-binding protein Musashi homolog 1; RNA-binding protein Musashi homolog 1 L+35bp; RNA-binding protein Musashi homolog 1 Long; RNA-binding protein Musashi homolog 1b; wu:fb40b12; zgc:103751; zMsi1
Host Species Mouse
Immunogen Full length human recombinant protein of human FAM40A produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362012, 490117, 85369
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.