Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSI1 Monoclonal Antibody (OTI3F4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502420

Description
Musashi-1 is a member of an evolutionarily conserved family of RNA-binding proteins that is prefentially expressed in the nervous system. It is a neural RNA-binding protein that is strongly expressed in fetal and adult neural stem cells (NSCs). Functionally, Musashi-1 plays important roles in cell fate decision, including the maintece of the stem-cell state, differentiation and tumorigenesis. Musashi-1 is an important regulator of the Notch-1 pathway.Specifications
MSI1 | |
Monoclonal | |
0.9 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O43347 | |
MSI1 | |
Full length human recombit protein of human MSI1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI3F4 | |
Unconjugated | |
MSI1 | |
musashi RNA binding protein 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4440 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction