Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSI1 Monoclonal Antibody (OTI3G4), TrueMAB™, OriGene
SDP

Catalog No. 501673432
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-3432 100 μL
50-254-1595 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-167-3432 Supplier OriGene Supplier No. TA502370
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Musashi-1 is a member of an evolutionarily conserved family of RNA-binding proteins that is prefentially expressed in the nervous system. It is a neural RNA-binding protein that is strongly expressed in fetal and adult neural stem cells (NSCs). Functionally, Musashi-1 plays important roles in cell fate decision, including the maintece of the stem-cell state, differentiation and tumorigenesis. Musashi-1 is an important regulator of the Notch-1 pathway.

Specifications

Antigen MSI1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3G4
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MSI1
Gene Accession No. O43347
Gene Alias musashi RNA binding protein 1
Gene Symbols MSI1
Host Species Mouse
Immunogen Full length human recombit protein of human MSI1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4440
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.