Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSI1 Monoclonal Antibody (OTI3G4), TrueMAB™, OriGene
SDP

Catalog No. 502541595
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1595 30 μL
50-167-3432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1595 Supplier OriGene Supplier No. TA502370S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MSI1 Monoclonal antibody specifically detects MSI1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MSI1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3G4
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PIP4K2A
Gene Accession No. P48426
Gene Alias fb40b12; hypothetical protein LOC541389; m-Msi-1; ms1; msi1; msi1b; Msi1h; Musahi1; musashi homolog 1; Musashi homolog 1(Drosophila); musashi RNA binding protein 1; musashi RNA binding protein 1b; musashi RNA-binding protein 1; musashi1; musashi-1; RNA-binding protein Musashi homolog 1; RNA-binding protein Musashi homolog 1 L+35bp; RNA-binding protein Musashi homolog 1 Long; RNA-binding protein Musashi homolog 1b; wu:fb40b12; zgc:103751; zMsi1
Host Species Mouse
Immunogen Full length human recombinant protein of human PIP4K2A produced in 293T Cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5305
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.