Learn More
MSI1 Mouse anti-Human, Clone: OTI2E9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502254

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Musashi-1 is a member of an evolutionarily conserved family of RNA-binding proteins that is prefentially expressed in the nervous system. It is a neural RNA-binding protein that is strongly expressed in fetal and adult neural stem cells (NSCs). Functionally, Musashi-1 plays important roles in cell fate decision, including the maintece of the stem-cell state, differentiation and tumorigenesis. Musashi-1 is an important regulator of the Notch-1 pathway.Specifications
MSI1 | |
Monoclonal | |
Unconjugated | |
MSI1 | |
musashi RNA binding protein 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4440 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2E9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O43347 | |
MSI1 | |
Full length human recombit protein of human MSI1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.