Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596187 Supplier Invitrogen™ Supplier No. PA596187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRVMPYGMDA FMLGIGMLGY PGFQATTYAS RSYTGLAPGY TYQFPEFRVE RTPLPSAPVL PELTAIPLTA YGPMAAAAAA AAVVRGTGSH PWTMAPPPGS TPSRTGGFLG TTSPGPMAEL YGAANQDSGV SSYISAASPA PSTGFGHSLG GPLIATAFTN GYH Positive Samples: A-549; Cellular Location: Cytoplasm, Nucleus.

Musashi-1 belongs to the Musashi family of RNA-binding proteins which play a role in cell fate determination, and maintenance of stem-cell state. Musashi family proteins are also involved in post transcriptional gene regulation. The gene encodes a protein with two conserved tandem RNA recognition motifs with highly conserved RNP (ribonucleoprotein) motifs. In mammals, Musashi-1 is expressed in fetal kidney, brain, liver and lung, and in adult brain and pancreas. In humans, the gene is located on chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSI1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene MSI1
Gene Accession No. O43347, Q61474, Q8K3P4
Gene Alias fb40b12; hypothetical protein LOC541389; m-Msi-1; ms1; msi1; msi1b; Msi1h; Musahi1; musashi homolog 1; Musashi homolog 1(Drosophila); musashi RNA binding protein 1; musashi RNA binding protein 1b; musashi RNA-binding protein 1; musashi1; musashi-1; RNA-binding protein Musashi homolog 1; RNA-binding protein Musashi homolog 1 L+35bp; RNA-binding protein Musashi homolog 1 Long; RNA-binding protein Musashi homolog 1b; wu:fb40b12; zgc:103751; zMsi1
Gene Symbols MSI1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-362 of human Musashi-1 (MSI1) (NP_0024331).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17690, 259272, 4440
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.