Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521145
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521145 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521145 Supplier Invitrogen™ Supplier No. PA521145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is EL4 cell lysate. PA5-21145 can be used with blocking peptide PEP-1259.

Msi2 (musashi homolog 2), also known as MSI2H, is a 328 amino acid protein that localizes to the cytoplasm and contains two RRM (RNA recognition motif) domains. Expressed ubiquitously at low levels, Msi2 functions as an RNA binding protein that, by regulating the expression of target mRNAs, is thought to play a role in the proliferation and maintenance of stem cells within the central nervous system. Msi2 is subject to posttranslational phosphorylation and is upregulated in response to brain injury, suggesting a role in healing and brain tissue regeneration. Chromosomal aberrations involving the Msi2 gene are associated with the progression of chronic myeloid leukemia. Multiple isoforms of Msi2 exist due to alternative splicing events.Tissue specificity: Ubiquitous; detected at low levels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSI2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MSI2
Gene Accession No. Q920Q6, Q96DH6
Gene Alias 1700105C15Rik; AI451722; AI563628; AW489193; MSI2; Msi2h; musashi homolog 2; musashi RNA binding protein 2; musashi RNA-binding protein 2; Musashi2; musashi-2; RGD1560397; RNA-binding protein Musashi homolog 2
Gene Symbols MSI2
Host Species Rabbit
Immunogen A 15 amino acid peptide near the amino terminus of human MSI2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124540, 360596, 76626
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.