Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MSK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536099
Description
This antibody detects endogenous protein at a molecular weight of 90 kDa. Purity is >95% by SDS-PAGE.
RPS6KA5 (MSK1) is a serine/threonine protein kinase that is essential for the regulation of various cellular processes. RPS6KA5 is required for the mitogen or stress-induced phosphorylation of the transcription factors CREB1 and ATF1 and for the regulation of the transcription factors RELA, STAT3 and ETV1/ER81, and that contributes to gene activation by histone phosphorylation and functions in the regulation of inflammatory genes. Phosphorylates CREB1 and ATF1 in response to mitogenic or stress stimuli such as UV-C irradiation, epidermal growth factor (EGF) and anisomycin. Plays an essential role in the control of RELA transcriptional activity in response to TNF and upon glucocorticoid, associates in the cytoplasm with the glucocorticoid receptor NR3C1 and contributes to RELA inhibition and repression of inflammatory gene expression. In skeletal myoblasts is required for phosphorylation of RELA at 'Ser-276' during oxidative stress. In erythropoietin-stimulated cells, is necessary for the 'Ser-727' phosphorylation of STAT3 and regulation of its transcriptional potential.
Specifications
MSK1 | |
Polyclonal | |
Unconjugated | |
Rps6ka5 | |
3110005L17Rik; 6330404E13Rik; 90 kDa ribosomal protein S6 kinase 5; AI854034; kinase MSK1; KS6A5; mitogen and stress-activated protein kinase 1; Msk1; MSPK1; Nuclear mitogen- and stress-activated protein kinase 1; ribosomal protein S6 kinase A5; ribosomal protein S6 kinase alpha-5; ribosomal protein S6 kinase, 90kDa, polypeptide 5; ribosomal protein S6 kinase, polypeptide 5; RLPK; RLSK; Rps6ka5; RSKL; RSK-like protein kinase; S6K-alpha-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
314384, 73086, 9252 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O75582, Q8C050 | |
Rps6ka5 | |
Synthetic peptide corresponding to amino acids 331-380 of human MSK1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction