Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSK1/RPS6KA5, p Ser376 Antibody (SR2276), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321542100UL
Description
MSK1/RPS6KA5 Monoclonal antibody specifically detects MSK1/RPS6KA5 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
MSK1/RPS6KA5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR2276 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RSKL, RSK-like protein kinase, S6K-alpha-5 | |
A synthesized peptide derived from human phospho-MSK1/RPS6KA5 [Ser376] (Uniprot #: O75582) | |
100 μg | |
Phospho Specific, Protein Kinase, Signal Transduction | |
9252 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction