Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582821 Supplier Invitrogen™ Supplier No. PA582821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQEVRNHPFF QGLDWVALAA RKIPAPFRPQ IRSELDVGNF AEEFTRLEPV YSPPGSPPPG DPRIFQGYSF VAPSILFDHN NAVMTDGLEA PGAGDRPGRA AVARSAMMQD SPFFQQYELD LREPALGQGS FSVC Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 95%.

RPS6KA4 (MSK2) is a serine/threonine protein kinase that is essential for the regulation of various cellular processes. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos. RPS6KA4 contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rps6ka4
Gene Accession No. O75676
Gene Alias 1110069D02Rik; 90 kDa ribosomal protein S6 kinase 4; 90kDa; AI848992; Ccdc88b; coiled-coil domain containing 88B; EC 2.7.11.1; kinase MSK2; KS6A4; mitogen- and stress-activated protein kinase 2; mitogen- and stress-activated protein kinase-2; mMSK2; MSK2; Nuclear mitogen- and stress-activated protein kinase 2; ribosomal protein kinase B; ribosomal protein S6 kinase A4; ribosomal protein S6 kinase alpha 4; ribosomal protein S6 kinase alpha-4; ribosomal protein S6 kinase, 90kD, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 4; ribosomal protein S6 kinase, polypeptide 4; RLSK; RPS6KA4; RSKB; RSK-B; RSK-like protein kinase; S6K-alpha-4
Gene Symbols Rps6ka4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPS6KA4. Recombinant protein control fragment (Product #RP-91470).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.