Learn More
Invitrogen™ MSL2 Monoclonal Antibody (3A4)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA522114
Catalog No. PIMA522114
Only null left
Add to Cart
Description
Description
Peptide Sequence: GQRCPCYSNR KACLDCICRG CQNSYMANGE KKLEAFAVPE KALEQTRLTL GINVTSIAVR NASTSTSVIN VTGSPVTTFL AASTHDDKSL DEAIDMRF.
Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at lysine 16 which is implicated in the formation of higher-order chromatin structure. Acts as an E3 ubiquitin ligase that promotes monoubiquitination of histone H2B at 'Lys-35' (H2BK34Ub), but not that of H2A. This activity is greatly enhanced by heterodimerization with MSL1. H2B ubiquitination in turn stimulates histone H3 methylation at 'Lys-4' (H3K4me) and 'Lys-79' (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSL2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9HCI7
MSL2
MSL2 (NP_060603, 478 a.a. ~ 575 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA
3A4
Unconjugated
MSL2
E130103E02Rik; E3 ubiquitin-protein ligase MSL2; E3 ubiquitin-protein transferase MSL2; Kiaa1585; male-specific lethal 2 homolog; male-specific lethal 2 homolog (Drosophila); male-specific lethal 2-like 1; Male-specific lethal-2 homolog; Male-specific lethal-2 homolog 1; MSL2; MSL-2; Msl2l1; MSL2-like 1; RGD1310355; ring finger protein 184; Rnf184
Mouse
Affinity chromatography
RUO
55167
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.