Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MSL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565518
Description
Immunogen sequence: DKAVKEKIPSH HFMPGSPTKT VYKKPQEKKG CKCGRATQNP SVLTCRGQRC PCYSNRKACL DCICRGCQNS YMANGEKKLE AFAVPEKALE QTRLTLGINV TSIAVRNAST STSVINVTGS PVTTFLAAST HDDKSLDEAI Highest antigen sequence identity to the following orthologs - mouse 99%, rat 100%.
Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at lysine 16 which is implicated in the formation of higher-order chromatin structure. Acts as an E3 ubiquitin ligase that promotes monoubiquitination of histone H2B at 'Lys-35' (H2BK34Ub), but not that of H2A. This activity is greatly enhanced by heterodimerization with MSL1. H2B ubiquitination in turn stimulates histone H3 methylation at 'Lys-4' (H3K4me) and 'Lys-79' (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1. [UniProt].
Specifications
MSL2 | |
Polyclonal | |
Unconjugated | |
MSL2 | |
E130103E02Rik; E3 ubiquitin-protein ligase MSL2; E3 ubiquitin-protein transferase MSL2; Kiaa1585; male-specific lethal 2 homolog; male-specific lethal 2 homolog (Drosophila); male-specific lethal 2-like 1; Male-specific lethal-2 homolog; Male-specific lethal-2 homolog 1; MSL2; MSL-2; Msl2l1; MSL2-like 1; RGD1310355; ring finger protein 184; Rnf184 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55167 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCI7 | |
MSL2 | |
Recombinant Human MSL2. Recombinant protein control fragment (Product #RP-104856). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction