Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588985 Supplier Thermo Scientific Supplier No. PA588985
Add to Cart
Add to Cart

MSL2 Polyclonal Antibody

MSL2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen MSL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MSL2
Gene Accession No. Q69ZF8, Q9HCI7
Gene Alias E130103E02Rik; E3 ubiquitin-protein ligase MSL2; E3 ubiquitin-protein transferase MSL2; Kiaa1585; male-specific lethal 2 homolog; male-specific lethal 2 homolog (Drosophila); male-specific lethal 2-like 1; Male-specific lethal-2 homolog; Male-specific lethal-2 homolog 1; MSL2; MSL-2; Msl2l1; MSL2-like 1; RGD1310355; ring finger protein 184; Rnf184
Gene Symbols MSL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 328-577 of human MSL2 (NP_0606032).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315959, 55167, 77853
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.