Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSL2L1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$610.00
Specifications
Antigen | MSL2L1 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, Immunoprecipitation, ChIP Assay |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MSL2L1 Polyclonal specifically detects MSL2L1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
MSL2L1 | |
Western Blot, Immunoprecipitation, ChIP Assay | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
55167 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SPEHSNTIDVCNTVDIKTEDLSDSLPPVCDTVATDLCSTGIDICSFSEDIKPGDSLLLSVEEVLRSLETVSNTEVCCPNLQPNLEATVSNGPFLQLSSQSLSHNVFMSTSPALHGLSCTAATPKIAKLNRKRSRSESDSEKVQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04-0.4 ug/ml, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
Polyclonal | |
Rabbit | |
Human | |
FLJ10546, KIAA1585FLJ54913, male-specific lethal 2 homolog, male-specific lethal 2 homolog (Drosophila), Male-specific lethal 2-like 1, male-specific lethal 2-like 1 (Drosophila), Male-specific lethal-2 homolog 1, MSL-2, MSL2L1, MSL2-like 1, PHNPCC4, PMS2, RING finger protein 184msl-2, RNF184 | |
MSL2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title