Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556967 Supplier Invitrogen™ Supplier No. PA556967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NPSTPQSTES QPTTGEPATP KRRKAEPEAL QSLRRSTRHS ANCDRLSESS ASPQPKRRQQ DTSASMPKLF LHLEKKTPVH SRSSSPIPLT PSKEGSAV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

This gene encodes a nuclear protein and has similarity to drosophila male-specific lethal-3 gene. The drosophila protein plays a critical role in a dosage-compensation pathway, which equalizes X-linked gene expression in males and females. Thus this encoded protein is thought to play a similar function in chromatin remodeling and transcriptional regulation. This gene has been found to undergo X inactivation. There are four alternatively spliced transcript variants of this gene encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Msl3
Gene Accession No. Q8N5Y2
Gene Alias AU018931; male-specific lethal 3 homolog; male-specific lethal 3 homolog (Drosophila); male-specific lethal-3 homolog 1; Male-specific lethal-3 protein-like 1; MSL3; Msl31; MSL3L1; MSL3-like 1
Gene Symbols Msl3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MSL3. Recombinant protein control fragment (Product #RP-95421).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.