Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSLN, Mouse anti-Human, Clone: MN-1, Abnova™

Catalog No. 89999093 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-093 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-093 Supplier Abnova Corporation Supplier No. MAB16418
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen MSLN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MN-1
Conjugate Unconjugated
Dilution ELISA (1:10000-1:50000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20 mM K3PO4, 150 mM NaCl, pH 7.2 (0.01% sodium azide).
Gene mesothelin
Gene Alias CAK1/MPF/SMR
Gene Symbols MSLN
Host Species Mouse
Immunogen Recombinant protein corresponding to the extracellular domain of human MSLN.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10232
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.