Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSLN Mouse anti-Human, Clone: OTI2E5, lyophilized, TrueMAB™
SDP

Catalog No. 501667307
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-7307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-7307 Supplier OriGene Supplier No. CF805172
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein that is present on the surface of normal mesothelium and is overexpressed in epithelial ovarian cancer and maligt mesotheliomas.

Specifications

Antigen MSLN
Applications Western Blot
Classification Monoclonal
Clone OTI2E5
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene MSLN
Gene Accession No. Q13421
Gene Alias MPF, SMRP
Gene Symbols MSLN
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 270-493 of human MSLN produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10232
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.