Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSLN Mouse anti-Human, Unconjugated, Clone: MSLN/2131, Abnova™

Catalog No. 89337357 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-357 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-357 Supplier Abnova Corporation Supplier No. MAB20860
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MSLN Monoclonal antibody specifically detects MSLN in Human samples. It is validated for IHC-P.

Specifications

Antigen MSLN
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MSLN/2131
Conjugate Unconjugated
Description mesothelin
Gene MSLN
Gene Alias CAK1, MPF, SMR
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 273-407 of human MSLN
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 10232
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.