Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MSMB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102533 Supplier Invitrogen™ Supplier No. PA5102533
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MSMB.

MSMB protein is a member of the immunoglobulin binding factor family. It is synthesized by the epithelial cells of the prostate gland and secreted into the seminal plasma. This protein has inhibin-like activity. It may have a role as an autocrine paracrine factor in uterine, breast and other female reproductive tissues. The expression of the encoded protein is found to be decreased in prostate cancer. Two alternatively spliced transcript variants encoding different isoforms are described for this gene. The use of alternate polyadenylation sites has been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSMB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MSMB
Gene Accession No. O08540, P08118
Gene Alias beta-inhibin; beta-microseminoprotein; beta-MSP; HPC 13; HPC13; IGBF; immunoglobulin binding factor; Immunoglobulin-binding factor; microseminoprotein beta; microseminoprotein, beta; microseminoprotein, beta-; Msmb; MSP; MSPB; PIP; PN 44; PN44; Prostate secreted seminal plasma protein; prostate secretory protein of 94 amino acids; prostate secretory protein PSP94; prostatic inhibin protein; PRPS; PRSP; PSP; PSP 57; PSP57; PSP94; PSP-94; Seminal plasma beta-inhibin
Gene Symbols MSMB
Host Species Rabbit
Immunogen A synthesized peptide derived from human MSMB(Accession P08118), corresponding to amino acid residues F24-S74.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17695, 4477
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.