Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MSOP
GREENER_CHOICE

Catalog No. 80325
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-325 25 mg
80-35 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 80-325 Supplier Tocris Bioscience™ Supplier No. 0803/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specific group III mGlu antagonist

MSOP is a selective group III metabotropic glutamate receptor antagonist. Displays an apparent KD of 51 μM for the L-AP4-sensitive presynaptic mGluR on primary afferent terminals in spinal cord compared to > 700 μM for the (1S,3S)-ACPD-sensitive presynaptic mGlu in the same system. Has no activity on postsynaptic mGlu receptors or on ionotropic glutamate receptors on neonatal rat motoneurons.

Specifications

Chemical Name or Material (RS)-α-Methylserine-O-phosphate
CAS 66515-29-5
Quantity 25 mg
Target Group III mGluR Antagonists
Molecular Formula C4H10NO6P
Formula Weight Observed MW: 199.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.