Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSP/MST1 Antibody (OTI1A10), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272792JF549
Description
MSP/MST1 Monoclonal specifically detects MSP/MST1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MSP/MST1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MST1 | |
Recombinant protein fragment corresponding to amino acids 194-462 of human MSP/MST1 (NP_066278) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1A10 | |
Western Blot, Immunohistochemistry | |
D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, NF15S2 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
4485.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction